Assay Details
| Gene Symbol: Tmprss11b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177024.4 | NP_795998.2 | 7 - 8 | 713 | 66 | |
| GenBank mRNA | AK036858.1 | - | 766 | 66 | ||
| AK036968.1 | - | 7 - 8 | 766 | 66 | ||
| AK137654.1 | - | 778 | 66 | |||
| BC115419.1 | - | 7 - 8 | 713 | 40093437 | 66 | |
| BC115420.2 | - | 7 - 8 | 713 | 40093438 | 66 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tmprss11b
Genomic Map