Assay Details
| Gene Symbol: Tmprss11b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177024.4 | NP_795998.2 | 9 - 10 | 1115 | 110 | |
| GenBank mRNA | AK036858.1 | - | 1168 | 110 | ||
| AK036968.1 | - | 9 - 10 | 1168 | 110 | ||
| AK137654.1 | - | 1180 | 110 | |||
| BC115419.1 | - | 9 - 10 | 1115 | 40093437 | 110 | |
| BC115420.2 | - | 9 - 10 | 1115 | 40093438 | 110 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2300421 | AK137654.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2062991 | BC115419.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2062991 | BC115420.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2062991 | AK036858.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2062991 | AK137654.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2062991 | NM_177024.4 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 108268 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 329783 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 755338 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 379146 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 588229 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 359097 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 229876 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 917106 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 865917 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 845987 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 129947 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 949352 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 64793 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 17003 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 35718 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P110993 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P135631 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2062991 | BC115419.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2062991 | BC115420.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2062991 | AK036858.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2062991 | AK137654.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2062991 | NM_177024.4 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2300421 | AK137654.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P329027 | AK036858.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P329027 | AK137654.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23864836 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23864835 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23864839 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23864841 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23864819 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23864818 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23864828 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23864826 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23864810 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23864822 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23864833 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23864820 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23864807 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23864832 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23864803 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23864838 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23864804 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23864824 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23864812 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23864811 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23864815 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23864823 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23864805 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23864799 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01153085 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P110993 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P135631 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05643445 | - | - |
| Whole Mouse Genome G4122A | A_51_P329027 | AK036858.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P329027 | AK137654.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P103577 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P135631 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P110993 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04350101 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04350096 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04350098 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04350092 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04350093 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04350095 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23864766 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23864800 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23864754 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23864675 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23864749 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23864803 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23864763 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23864794 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23864845 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23864785 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23864807 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23864818 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23864681 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23864755 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23864758 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23864772 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23864835 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23864762 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23864713 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23864756 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23864773 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23864686 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23864792 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23864757 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23864777 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23864810 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23864799 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23864715 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23864708 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23864736 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23864839 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23864716 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23864815 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23864712 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23864823 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23864723 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23864709 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23864727 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23864767 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23864692 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23864679 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23864729 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23864726 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23864781 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23864740 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23864796 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23864752 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23864832 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23864779 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23864760 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23864678 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23864732 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23864822 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23864700 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23864737 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23864829 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23864844 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23864804 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23864847 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23864770 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23864826 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23864833 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23864739 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23864820 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23864697 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23864691 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23864731 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23864672 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23864744 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23864812 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23864788 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23864774 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23864848 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23864718 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23864694 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23864719 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23864687 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23864721 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23864747 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23864786 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23864769 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23864676 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23864741 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23864730 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23864693 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23864819 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23864804 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23864786 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23864823 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23864794 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23864799 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23864820 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23864784 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23864807 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04425133 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23864822 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23864781 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23864819 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23864785 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23864815 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23864777 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23864775 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23864824 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04350104 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04905085 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23864779 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23864810 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23864812 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23864837 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23864793 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23864843 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04668322 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23864829 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23864792 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04151352 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23864800 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23864833 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23864841 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23864788 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23864826 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23864803 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23864778 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23864796 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23864832 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10531018 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10531015 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10531013 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10531020 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10531019 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10531011 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10531012 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10531014 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10531017 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10531016 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10531009 | - | - |
| Affymetrix Mouse Clariom S Array | PSR0500023709.mm.1 | NM_177024.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500023709.mm.1 | NM_177024.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500023709.mm.1 | TC0500002685.mm.1 | MTA TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 630301 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0000630301 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2778024 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2778024 | - | - |
| mouse-6 v1.1 (Illumina) | scl26426.10_687-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 630301 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl26426.10_687-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 630301 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2882189 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2882189 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2778024 | - | - |

Tmprss11b
Genomic Map