Assay Details
| Gene Symbol: Flad1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177041.3 | NP_796015.2 | 1 - 2 | 78 | 80 | |
| GenBank mRNA | AK044501.1 | - | 87 | 80 | ||
| BC046769.1 | - | 1 - 2 | 78 | 6809196 | 80 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK044501.1, BC046769.1, NM_177041.3 transcripts.
Set Membership: |
|

Flad1
Genomic Map