Assay Details
| Gene Symbol: Flad1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177041.3 | NP_796015.2 | 3 - 4 | 1044 | 66 | |
| GenBank mRNA | AK044501.1 | - | 1053 | 66 | ||
| AK161929.1 | - | 3 - 4 | 1088 | 66 | ||
| BC025817.1 | - | 3 - 4 | 997 | 5099127 | 66 | |
| BC046769.1 | - | 3 - 4 | 1044 | 6809196 | 66 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2107025 | AK044501.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2107025 | BC006806.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2056140 | BC046769.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2056140 | AK161929.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2056140 | BC025817.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2056140 | NM_177041.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2056140 | AK044501.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_65_P11193 | BC046769.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_65_P11193 | AK161929.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_65_P11193 | BC025817.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_65_P11193 | NM_177041.3 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 67054 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 775604 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 226100 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 617696 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 696884 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 403970 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 169634 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 44169 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2107025 | AK044501.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2107025 | BC006806.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2056140 | BC046769.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2056140 | AK161929.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2056140 | BC025817.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2056140 | NM_177041.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2056140 | AK044501.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_65_P11193 | BC046769.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_65_P11193 | AK161929.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_65_P11193 | BC025817.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_65_P11193 | NM_177041.3 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22319242 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22319232 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22319237 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22319225 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22319231 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22319241 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22319234 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22319239 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22319226 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22319223 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22319235 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22319229 | - | - |
| Whole Mouse Genome G4122A | A_52_P356703 | BC046769.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P356703 | AK161929.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P356703 | BC025817.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P356703 | NM_177041.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P356703 | AK044501.1 | mouse_transcriptome |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02904682 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02904687 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02813005 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22319184 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22319274 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22319172 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22319262 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22319128 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22319229 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22319263 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22319155 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22319250 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22319152 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22319143 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22319137 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22319276 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22319133 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22319225 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22319160 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22319109 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22319125 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22319132 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22319272 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22319181 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22319279 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22319273 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22319105 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22319178 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22319117 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22319284 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22319146 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22319158 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22319170 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22319267 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22319102 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22319147 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22319106 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22319162 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22319103 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22319282 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22319129 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22319280 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22319185 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22319265 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22319269 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22319258 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22319232 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22319131 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22319260 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22319142 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22319123 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22319120 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22319108 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22319241 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22319139 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22319111 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22319242 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22319175 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22319183 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22319159 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22319115 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22319249 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22319223 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22319168 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22319153 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22319270 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22319135 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22319235 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22319232 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02707538 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22319237 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02813005 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02843910 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02904682 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22319229 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02843909 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22319226 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02904687 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22319240 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02843914 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22319234 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10499638 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10499627 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10499635 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10499634 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10499629 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10499637 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10499633 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10499626 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10499630 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10499628 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10499632 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10499631 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1424421_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10499620 | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 319945_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 319945_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0300018127.mm.1 | NM_177041.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300018127.mm.1 | NM_177041.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300018127.mm.1 | TC0300002351.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0300018126.mm.1 | NM_177041.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300018126.mm.1 | NM_177041.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300018126.mm.1 | TC0300002351.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1424422_s_at | - | - |
| Mouse Expression Set 430 | 1424423_PM_at | - | - |
| Mouse Expression Set 430 | 1424422_PM_s_at | - | - |
| Mouse Expression Set 430 | 1424423_at | - | - |
| Mouse Genome 430A 2.0 Array | 1424423_at | - | - |
| Mouse Genome 430A 2.0 Array | 1424423_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1424422_PM_s_at | - | - |
| Mouse Genome 430A 2.0 Array | 1424422_s_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1424421_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1424421_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 642619 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 4670458 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 4670458 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 2630563 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002630563 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004670458 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2602630 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2602630 | - | - |
| mouse-6 v1.1 (Illumina) | scl00319945.1_316-S | - | - |
| mouse-6 v1.1 (Illumina) | scl0319945.1_51-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2630563 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4670458 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl00319945.1_316-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0319945.1_51-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2630563 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4670458 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2602630 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1252499 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1252499 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2602630 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl0319945.1_51-S | - | - |

Flad1
Genomic Map