Assay Details
| Gene Symbol: Fstl4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177059.3 | NP_796033.2 | 14 - 15 | 1895 | 144 | |
| GenBank mRNA | AF374459.1 | - | 14 - 15 | 1895 | 144 | |
| AK046350.1 | - | 14 - 15 | 1946 | 144 | ||
| AK081114.1 | - | 14 - 15 | 1941 | 144 | ||
| AK220367.1 | - | 10 - 11 | 1275 | 144 | ||
| BC132353.1 | - | 14 - 15 | 1872 | 40130630 | 144 | |
| BC144824.1 | - | 14 - 15 | 1865 | 144 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Fstl4
Genomic Map