Assay Details
| Gene Symbol: Fstl4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177059.3 | NP_796033.2 | 1 - 2 | 175 | 81 | |
| GenBank mRNA | AF374459.1 | - | 1 - 2 | 175 | 81 | |
| AK046350.1 | - | 1 - 2 | 225 | 81 | ||
| AK081114.1 | - | 1 - 2 | 225 | 81 | ||
| BC132353.1 | - | 1 - 2 | 152 | 40130630 | 81 | |
| BC144824.1 | - | 1 - 2 | 152 | 81 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AF374459.1, AK046350.1, AK081114.1, BC132353.1, NM_177059.3 transcripts.
Set Membership: |
|

Fstl4
Genomic Map