Assay Details
| Gene Symbol: Fstl4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177059.3 | NP_796033.2 | 7 - 8 | 1082 | 93 | |
| GenBank mRNA | AF374459.1 | - | 7 - 8 | 1082 | 93 | |
| AK046350.1 | - | 7 - 8 | 1133 | 93 | ||
| AK081114.1 | - | 7 - 8 | 1132 | 93 | ||
| AK220367.1 | - | 3 - 4 | 462 | 93 | ||
| BC132353.1 | - | 7 - 8 | 1059 | 40130630 | 93 | |
| BC144824.1 | - | 7 - 8 | 1059 | 93 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK046350.1 transcripts.
Set Membership: |
|

Fstl4
Genomic Map