Assay Details
| Gene Symbol: Fstl4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177059.3 | NP_796033.2 | 9 - 10 | 1366 | 69 | |
| GenBank mRNA | AF374459.1 | - | 9 - 10 | 1366 | 69 | |
| AK046350.1 | - | 9 - 10 | 1417 | 69 | ||
| AK081114.1 | - | 9 - 10 | 1416 | 69 | ||
| AK220367.1 | - | 5 - 6 | 746 | 69 | ||
| BC132353.1 | - | 9 - 10 | 1343 | 40130630 | 69 | |
| BC144824.1 | - | 9 - 10 | 1336 | 69 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Fstl4
Genomic Map