Assay Details
| Gene Symbol: Gnal | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010307.3 | NP_034437.1 | 2 - 3 | 488 | 93 | |
| NM_177137.5 | NP_796111.2 | 2 - 3 | 653 | 93 | ||
| GenBank mRNA | AK032407.1 | - | 2 - 3 | 299 | 93 | |
| AK035952.1 | - | 2 - 3 | 654 | 93 | ||
| AK045484.1 | - | 2 - 3 | 618 | 93 | ||
| AK158831.1 | - | 2 - 3 | 489 | 93 | ||
| AY179169.1 | - | 2 - 3 | 222 | 93 | ||
| BC078439.1 | - | 2 - 3 | 615 | 5685967 | 93 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gnal
Genomic Map