Assay Details
| Gene Symbol: Enpp6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177304.3 | NP_796278.1 | 4 - 5 | 721 | 92 | |
| GenBank mRNA | AK038460.1 | - | 4 - 5 | 693 | 92 | |
| AK046797.1 | - | 4 - 5 | 722 | 92 | ||
| AK046881.1 | - | 4 - 5 | 726 | 92 | ||
| AK079642.1 | - | 4 - 5 | 721 | 92 | ||
| BC096376.1 | - | 4 - 5 | 692 | 4237600 | 92 | |
More Information
Updated Mapping Information
Set Membership: |
|

Enpp6
Genomic Map