Assay Details
| Gene Symbol: Slfnl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177570.3 | NP_808238.1 | 3 - 4 | 1355 | 80 | |
| GenBank mRNA | AK077106.1 | - | 3 - 4 | 1356 | 80 | |
| BC117545.1 | - | 2 - 3 | 1103 | 40105640 | 80 | |
| BC118052.1 | - | 2 - 3 | 1106 | 40105639 | 80 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slfnl1
Genomic Map