Assay Details
| Gene Symbol: Gm38404 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_073373.1 | 3 - 4 | 566 | 69 | ||
| GenBank mRNA | AK051577.1 | - | 3 - 4 | 567 | 69 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | RIKEN cDNA 9330179D12 gene | 9330179D12Rik | -83.8kb | Anti-sense | Chr. 6 - 127149389 - 127212419 on NCBI Build GRCm38 | |
| Downstream |
poly (ADP-ribose) polymerase family, member 11 |
Parp11 |
5330431N24Rik, AI851877, ARTD11, HIN1L |
148.5kb | Anti-sense | Chr. 6 - 127446845 - 127512908 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Gm38404
Genomic Map