Assay Details
| Gene Symbol: Fhod1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177699.4 | NP_808367.2 | 4 - 5 | 558 | 76 | |
| GenBank mRNA | AB041045.1 | - | 4 - 5 | 452 | 76 | |
| AK030737.1 | - | 4 - 5 | 520 | 76 | ||
| BC060654.1 | - | 4 - 5 | 541 | 30354865 | 76 | |
More Information
Updated Mapping Information
Set Membership: |
|

Fhod1
Genomic Map