Assay Details
| Gene Symbol: Mcmdc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177722.3 | NP_808390.2 | 5 - 6 | 571 | 67 | |
| GenBank mRNA | AK031395.1 | - | 5 - 6 | 529 | 67 | |
| CA978963.1 | - | 3 - 4 | 326 | 67 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mcmdc2
Genomic Map