Assay Details
| Gene Symbol: Fat4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_183221.3 | NP_899044.3 | 2 - 3 | 5324 | 106 | |
| GenBank mRNA | AK034552.1 | - | 2 - 3 | 1322 | 106 | |
| DQ286572.1 | - | 2 - 3 | 5324 | 106 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Fat4
Genomic Map