Assay Details
| Gene Symbol: E2f2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK087452.1 | - | 5 - 6 | 1244 | 76 | |
| AK157322.1 | - | 2 - 3 | 830 | 76 | ||
| AK157676.1 | - | 3 - 4 | 394 | 76 | ||
| BC062101.1 | - | 5 - 6 | 951 | 6848237 | 76 | |
More Information
Updated Mapping Information
Set Membership: |
|

E2f2
Genomic Map