Assay Details
| Gene Symbol: E2f2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK087452.1 | - | 6 - 7 | 1441 | 62 | |
| BC062101.1 | - | 6 - 7 | 1148 | 6848237 | 62 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: E2f2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK087452.1 | - | 6 - 7 | 1441 | 62 | |
| BC062101.1 | - | 6 - 7 | 1148 | 6848237 | 62 | |
Set Membership: |
|