Assay Details
| Gene Symbol: Ogfod1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001093757.1 | NP_001087226.1 | 7 - 8 | 864 | 142 | |
| NM_177767.4 | NP_808435.3 | 8 - 9 | 993 | 142 | ||
| GenBank mRNA | AK029629.1 | - | 8 - 9 | 995 | 142 | |
| AK122532.1 | - | 8 - 9 | 897 | 142 | ||
| AK156765.1 | - | 8 - 9 | 963 | 142 | ||
| AK164189.1 | - | 7 - 8 | 751 | 142 | ||
| AK166343.1 | - | 8 - 9 | 916 | 142 | ||
| BC040267.1 | - | 7 - 8 | 824 | 5364674 | 142 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ogfod1
Genomic Map