Assay Details
| Gene Symbol: Fa2h | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178086.3 | NP_835187.2 | 5 - 6 | 856 | 128 | |
| GenBank mRNA | AK033736.1 | - | 5 - 6 | 854 | 128 | |
| AK090338.1 | - | 5 - 6 | 850 | 128 | ||
| AK161502.1 | - | 5 - 6 | 854 | 128 | ||
| AY660882.1 | - | 5 - 6 | 787 | 128 | ||
| BC026629.1 | - | 3 - 4 | 398 | 128 | ||
| BC046985.1 | - | 5 - 6 | 812 | 128 | ||
| BC111912.1 | - | 6 - 7 | 677 | 128 | ||
| BC128080.1 | - | 5 - 6 | 837 | 40104374 | 128 | |
| BC128081.1 | - | 5 - 6 | 837 | 40104375 | 128 | |
More Information
Updated Mapping Information
Set Membership: |
|

Fa2h
Genomic Map