Assay Details
| Gene Symbol: Trim62 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178110.2 | NP_835211.1 | 1 - 2 | 434 | 64 | |
| GenBank mRNA | AK186103.1 | - | 1 - 2 | 63 | ||
| AK189353.1 | - | 1 - 2 | 63 | |||
| BC049095.1 | - | 1 - 2 | 434 | 6416690 | 64 | |
More Information
Updated Mapping Information
Set Membership: |
|

Trim62
Genomic Map