Assay Details
| Gene Symbol: Agmo | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178767.5 | NP_848882.2 | 6 - 7 | 1078 | 146 | |
| GenBank mRNA | AK039191.1 | - | 6 - 7 | 649 | 146 | |
| AK040702.1 | - | 6 - 7 | 1079 | 146 | ||
| AK050246.1 | - | 6 - 7 | 939 | 146 | ||
| AK163155.1 | - | 6 - 7 | 859 | 146 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Agmo
Genomic Map