Assay Details
| Gene Symbol: Nceh1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178772.3 | NP_848887.1 | 1 - 2 | 207 | 66 | |
| GenBank mRNA | AK034339.1 | - | 1 - 2 | 134 | 66 | |
| AK038691.1 | - | 208 | 66 | |||
| AK045363.1 | - | 1 - 2 | 208 | 66 | ||
| AK085767.1 | - | 1 - 2 | 208 | 66 | ||
| AK128968.1 | - | 1 - 2 | 218 | 66 | ||
| AK135837.1 | - | 1 - 2 | 197 | 66 | ||
| AK173158.1 | - | 1 - 2 | 177 | 66 | ||
| BC082569.1 | - | 1 - 2 | 190 | 30550100 | 66 | |
More Information
Updated Mapping Information
Set Membership: |
|

Nceh1
Genomic Map