Assay Details
| Gene Symbol: Prkd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252458.1 | NP_001239387.1 | 4 - 5 | 782 | 100 | |
| NM_178900.4 | NP_849231.1 | 3 - 4 | 1067 | 100 | ||
| GenBank mRNA | AK037030.1 | - | 3 - 4 | 1068 | 100 | |
| AK041207.1 | - | 2 - 3 | 287 | 100 | ||
| AK154029.1 | - | 3 - 4 | 916 | 100 | ||
| AK170236.1 | - | 4 - 5 | 755 | 100 | ||
| AK170761.1 | - | 4 - 5 | 746 | 100 | ||
| BC095949.1 | - | 4 - 5 | 732 | 4950957 | 100 | |
| BC096444.1 | - | 4 - 5 | 750 | 4216827 | 100 | |
More Information
Updated Mapping Information
Set Membership: |
|

Prkd2
Genomic Map