Assay Details
| Gene Symbol: Prkd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252458.1 | NP_001239387.1 | 7 - 8 | 1242 | 78 | |
| NM_178900.4 | NP_849231.1 | 6 - 7 | 1527 | 78 | ||
| GenBank mRNA | AK037030.1 | - | 6 - 7 | 1528 | 78 | |
| AK041207.1 | - | 5 - 6 | 747 | 78 | ||
| AK170761.1 | - | 7 - 8 | 1206 | 78 | ||
| BC095949.1 | - | 7 - 8 | 1192 | 4950957 | 78 | |
| BC096444.1 | - | 7 - 8 | 1210 | 4216827 | 78 | |
More Information
Updated Mapping Information
Set Membership: |
|

Prkd2
Genomic Map