Assay Details
| Gene Symbol: Prkd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252458.1 | NP_001239387.1 | 10 - 11 | 1596 | 79 | |
| NM_178900.4 | NP_849231.1 | 9 - 10 | 1881 | 79 | ||
| GenBank mRNA | AK037030.1 | - | 9 - 10 | 1882 | 79 | |
| AK154029.1 | - | 9 - 10 | 1730 | 79 | ||
| AK170236.1 | - | 10 - 11 | 1569 | 79 | ||
| AK170761.1 | - | 10 - 11 | 1560 | 79 | ||
| BC095949.1 | - | 10 - 11 | 1546 | 4950957 | 79 | |
| BC096444.1 | - | 10 - 11 | 1564 | 4216827 | 79 | |
More Information
Updated Mapping Information
Set Membership: |
|

Prkd2
Genomic Map