Assay Details
| Gene Symbol: Atf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007498.3 | NP_031524.2 | 1 - 2 | 236 | 95 | |
| GenBank mRNA | AK133965.1 | - | 1 - 2 | 240 | 95 | |
| AK156786.1 | - | 1 - 2 | 156 | 95 | ||
| BC019946.1 | - | 1 - 2 | 247 | 4011514 | 95 | |
| BC064799.1 | - | 1 - 2 | 102 | 30054998 | 95 | |
| U19118.1 | - | 1 - 2 | 158 | 95 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK133965.1, AK156786.1, BC019946.1, BC064799.1, NM_007498.3, U19118.1 transcripts.
Set Membership: |
|

Atf3
Genomic Map