Assay Details
| Gene Symbol: Bid | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007544.3 | NP_031570.2 | 5 - 6 | 720 | 118 | |
| GenBank mRNA | AK045731.1 | - | 5 - 6 | 735 | 118 | |
| AK051076.1 | - | 736 | 119 | |||
| AK052356.1 | - | 5 - 6 | 725 | 118 | ||
| AK077657.1 | - | 5 - 6 | 703 | 118 | ||
| AK161235.1 | - | 5 - 6 | 742 | 118 | ||
| BC002031.1 | - | 5 - 6 | 720 | 3486537 | 118 | |
| U75506.1 | - | 4 - 5 | 597 | 118 | ||
More Information
Set Membership: |
|

Bid
Genomic Map