Assay Details
| Gene Symbol: Dnajc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008929.3 | NP_032955.2 | 7 - 8 | 999 | 123 | |
| GenBank mRNA | AK004033.1 | - | 2 - 3 | 300 | 123 | |
| AK146097.1 | - | 7 - 8 | 963 | 123 | ||
| AK148270.1 | - | 7 - 8 | 956 | 123 | ||
| BC013766.1 | - | 7 - 8 | 944 | 2646147 | 123 | |
| U28423.1 | - | 7 - 8 | 942 | 123 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Dnajc3
Genomic Map