Assay Details
| Gene Symbol: Zfp185 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109043.1 | NP_001102513.1 | 14 - 15 | 1328 | 129 | |
| NM_009549.3 | NP_033575.3 | 14 - 15 | 1331 | 129 | ||
| GenBank mRNA | AK014497.1 | - | 3 - 4 | 379 | 129 | |
| AK048188.1 | - | 14 - 15 | 1309 | 129 | ||
| BC039772.1 | - | 11 - 12 | 916 | 4191053 | 129 | |
| EF494744.1 | - | 14 - 15 | 1311 | 129 | ||
| U46687.1 | - | 13 - 14 | 1157 | 129 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Zfp185
Genomic Map