Assay Details
| Gene Symbol: Efnb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010111.5 | NP_034241.2 | 3 - 4 | 674 | 73 | |
| GenBank mRNA | AK132512.1 | - | 3 - 4 | 1141 | 73 | |
| AK132845.1 | - | 3 - 4 | 967 | 73 | ||
| AK134108.1 | - | 3 - 4 | 1141 | 73 | ||
| AK142203.1 | - | 3 - 4 | 1144 | 73 | ||
| AK142283.1 | - | 3 - 4 | 1144 | 73 | ||
| AK143059.1 | - | 3 - 4 | 1141 | 73 | ||
| BC057009.1 | - | 3 - 4 | 962 | 6827408 | 73 | |
| CT010381.1 | - | 3 - 4 | 515 | 73 | ||
| L38847.1 | - | 3 - 4 | 675 | 73 | ||
| U30244.1 | - | 3 - 4 | 512 | 73 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Efnb2
Genomic Map