Assay Details
| Gene Symbol: Gypa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC148192.1 | - | 1 - 2 | 307 | 40045496 | 121 |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC148192.1 transcripts.
Set Membership: |
|

Gypa
Genomic Map