Assay Details
| Gene Symbol: Lamp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017959.2 | NP_001017959.1 | 3 - 4 | 534 | 152 | |
| NM_001290485.1 | NP_001277414.1 | 3 - 4 | 534 | 152 | ||
| NM_010685.4 | NP_034815.2 | 3 - 4 | 534 | 152 | ||
| GenBank mRNA | AK012149.1 | - | 3 - 4 | 493 | 152 | |
| AK032974.1 | - | 3 - 4 | 496 | 152 | ||
| AK048174.1 | - | 3 - 4 | 476 | 152 | ||
| AK051989.1 | - | 2 - 3 | 1332 | 152 | ||
| AK078193.1 | - | 3 - 4 | 528 | 152 | ||
| AK088804.1 | - | 4 - 5 | 511 | 152 | ||
| AK128946.1 | - | 1 - 2 | 196 | 152 | ||
| AK136550.1 | - | 3 - 4 | 495 | 151 | ||
| AK146731.1 | - | 3 - 4 | 515 | 152 | ||
| AK150766.1 | - | 3 - 4 | 503 | 152 | ||
| AK159272.1 | - | 3 - 4 | 489 | 152 | ||
| AK163933.1 | - | 3 - 4 | 535 | 152 | ||
| AK217258.1 | - | 1 - 2 | 143 | 151 | ||
| BC138718.1 | - | 3 - 4 | 421 | 8861740 | 152 | |
| BC138723.1 | - | 3 - 4 | 421 | 8861745 | 152 | |
| J05287.1 | - | 3 - 4 | 461 | 152 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Lamp2
Genomic Map