Assay Details
| Gene Symbol: Lamp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010685.4 | NP_034815.2 | 8 - 9 | 1262 | 100 | |
| GenBank mRNA | AK012149.1 | - | 8 - 9 | 1221 | 100 | |
| AK032974.1 | - | 8 - 9 | 1224 | 100 | ||
| AK048174.1 | - | 8 - 9 | 1204 | 100 | ||
| AK088804.1 | - | 9 - 10 | 1239 | 100 | ||
| AK128946.1 | - | 6 - 7 | 924 | 100 | ||
| AK136550.1 | - | 8 - 9 | 1223 | 100 | ||
| AK163933.1 | - | 8 - 9 | 1263 | 100 | ||
| BC138718.1 | - | 8 - 9 | 1149 | 8861740 | 100 | |
| BC138723.1 | - | 8 - 9 | 1149 | 8861745 | 100 | |
More Information
Updated Mapping Information
Set Membership: |
|

Lamp2
Genomic Map