Assay Details
| Gene Symbol: Xcr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011798.4 | NP_035928.2 | 1 - 2 | 85 | 64 | |
| GenBank mRNA | AK040878.1 | - | 1 - 2 | 86 | 64 | |
| AK089908.1 | - | 1 - 2 | 86 | 64 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK040878.1, AK089908.1, NM_011798.4 transcripts.
Set Membership: |
|

Xcr1
Genomic Map