Assay Details
| Gene Symbol: Tmem45a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019631.3 | NP_062605.2 | 4 - 5 | 942 | 77 | |
| GenBank mRNA | AK049837.1 | - | 4 - 5 | 943 | 77 | |
| AK081478.1 | - | 4 - 5 | 946 | 77 | ||
| BC064715.1 | - | 4 - 5 | 945 | 30246737 | 77 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tmem45a
Genomic Map