Assay Details
| Gene Symbol: Atg5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053069.5 | NP_444299.1 | 1 - 2 | 226 | 135 | |
| GenBank mRNA | AK028315.1 | - | 1 - 2 | 228 | 135 | |
| BC002166.1 | - | 1 - 2 | 92 | 3486093 | 135 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK028315.1, BC002166.1, NM_053069.5 transcripts.
Set Membership: |
|

Atg5
Genomic Map