Assay Details
| Gene Symbol: Cerk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145475.4 | NP_663450.3 | 7 - 8 | 1287 | 62 | |
| GenBank mRNA | AK042077.1 | - | 7 - 8 | 878 | 62 | |
| AK052269.1 | - | 7 - 8 | 1053 | 62 | ||
| AK161801.1 | - | 125 | 62 | |||
| AK169528.1 | - | 7 - 8 | 872 | 62 | ||
| BC094253.1 | - | 7 - 8 | 1286 | 6400872 | 62 | |
More Information
Set Membership: |
|

Cerk
Genomic Map