Assay Details
| Gene Symbol: Tmc7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172476.4 | NP_766064.2 | 10 - 11 | 1467 | 62 | |
| GenBank mRNA | AK082562.1 | - | 10 - 11 | 1496 | 62 | |
| AK083189.1 | - | 10 - 11 | 1470 | 62 | ||
| AK132820.1 | - | 10 - 11 | 1497 | 62 | ||
| AY236499.1 | - | 10 - 11 | 1528 | 62 | ||
| AY263161.1 | - | 10 - 11 | 1496 | 62 | ||
| BC105966.1 | - | 4 - 5 | 498 | 62 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tmc7
Genomic Map