Assay Details
| Gene Symbol: Zer1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290503.1 | NP_001277432.1 | 7 - 8 | 1598 | 97 | |
| NM_178694.4 | NP_848809.2 | 7 - 8 | 1637 | 97 | ||
| GenBank mRNA | AK029290.1 | - | 7 - 8 | 1572 | 97 | |
| AK032153.1 | - | 6 - 7 | 1666 | 97 | ||
| AK032318.1 | - | 6 - 7 | 1244 | 97 | ||
| AK048853.1 | - | 7 - 8 | 1493 | 97 | ||
| AK081355.1 | - | 7 - 8 | 1475 | 97 | ||
| AK087046.1 | - | 7 - 8 | 1471 | 97 | ||
| AK155401.1 | - | 7 - 8 | 1545 | 97 | ||
| BC048924.1 | - | 7 - 8 | 1480 | 6312691 | 97 | |
| BC057021.1 | - | 7 - 8 | 1542 | 6825937 | 97 | |
More Information
Updated Mapping Information
Set Membership: |
|

Zer1
Genomic Map