Assay Details
| Gene Symbol: Zer1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290503.1 | NP_001277432.1 | 9 - 10 | 1987 | 67 | |
| NM_178694.4 | NP_848809.2 | 9 - 10 | 2026 | 67 | ||
| GenBank mRNA | AK155401.1 | - | 9 - 10 | 1934 | 67 | |
| BC048924.1 | - | 9 - 10 | 1869 | 6312691 | 67 | |
| BC057021.1 | - | 9 - 10 | 1931 | 6825937 | 67 | |
More Information
Updated Mapping Information
Set Membership: |
|

Zer1
Genomic Map