Assay Details
| Gene Symbol: Defa4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC134360.1 | - | 183 | 40134597 | 125 | |
| BX634116.1 | - | 1 - 2 | 125 | |||
More Information
Updated Mapping Information
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P497255 | NM_010039.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P497255 | BC134360.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P497255 | NM_010039.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P497255 | BC134360.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P994399 | NM_001177522.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P994399 | NM_001177481.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06444925 | - | - |
| Whole Mouse Genome G4122A | A_52_P994399 | NM_001177522.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P994399 | NM_001177481.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P138772 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P127364 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P06444925 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10570733 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10570661 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10577423 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10570718 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1422422_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10570732 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10570660 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10570717 | - | - |
| Mouse Genome 430 2.0 Array | 1422422_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1422422_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0800001798.mm.1 | NM_001177481.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800001798.mm.1 | NM_001177481.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800001798.mm.1 | TC0800000213.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1422422_PM_at | - | - |
| Mouse Expression Set 430 | 1422422_at | - | - |
| Murine Genome U74v2 Set | 92812_f_at | - | - |
| Murine Genome U74v2 Set | 102814_f_at | - | - |
| Murine Genome U74v2 Set | 102814_PM_f_at | - | - |
| Murine Genome U74v2 Set | 92812_PM_f_at | - | - |
| Mouse Genome 430A 2.0 Array | 1422422_at | - | - |
| Mouse Genome 430A 2.0 Array | 1422422_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1422422_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1422422_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 893404 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 6040390 | - | - |
| MouseWG-6 v2 Expression BeadChip | 100130280 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 100130280 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 6040390 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006040390 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2710914 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2710914 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2524846 | - | - |
| mouse-6 v1.1 (Illumina) | scl0013238.1_204-S | - | - |
| mouse-6 v1.1 (Illumina) | GI_6753625-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6040390 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 100130280 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0013238.1_204-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 100130280 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6040390 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2710914 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2524846 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2710914 | - | - |

Defa4
Genomic Map