Assay Details
| Gene Symbol: Olfr351 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146942.1 | NP_667153.1 | 1 | 664 | 117 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1952945 | NM_146942.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2102941 | NM_146942.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P146305 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2102941 | NM_146942.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1952945 | NM_146942.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P441120 | NM_146942.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21140311 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21140305 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21140308 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21140309 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21140302 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21140303 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21140304 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21140313 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21140299 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21140297 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P146305 | - | - |
| Whole Mouse Genome G4122A | A_51_P441120 | NM_146942.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P146305 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01231346 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01231348 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01231353 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01231369 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01231371 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21140296 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21140284 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21140303 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21140236 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21140250 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21140309 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21140311 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21140256 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21140305 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21140270 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21140247 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21140251 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21140283 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21140313 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21140281 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21140273 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21140231 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21140234 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21140279 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21140300 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21140294 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21140275 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21140232 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21140223 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21140245 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21140287 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21140243 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21140319 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21140297 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21140276 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21140290 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21140238 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21140225 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21140254 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21140242 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21140261 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21140241 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21140268 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21140283 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21140254 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21140270 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21140276 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21140290 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21140252 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01174075 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21140275 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01174082 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01389567 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21140279 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21140256 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01946423 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01174059 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21140284 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01174081 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21140294 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21140273 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21140258 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21140287 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10375792 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10482124 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10482123 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10375791 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 4760403 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 4760403 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004760403 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2665824 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2665824 | - | - |
| mouse-6 v1.1 (Illumina) | scl4525.1.1_51-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4760403 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl4525.1.1_51-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4760403 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2665824 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2665824 | - | - |

Olfr351
Genomic Map