Assay Details
| Gene Symbol: Tcl1b4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013774.2 | NP_038802.1 | 3 - 4 | 451 | 129 | |
| GenBank mRNA | AF195492.1 | - | 3 - 4 | 451 | 129 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2112982 | BC145788.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2112982 | BC145148.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2112982 | NM_013774.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2112982 | AF195492.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2112982 | AK139906.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P331809 | BC145788.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P331809 | BC145148.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P331809 | NM_013774.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P331809 | AF195492.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P331809 | AK139906.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P331809 | BC145788.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P331809 | BC145148.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P331809 | NM_013774.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P331809 | AF195492.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P331809 | AK139906.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2014626 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 342434 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 658676 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 109631 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P331809 | BC145788.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P331809 | BC145148.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P331809 | NM_013774.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P331809 | AF195492.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P331809 | AK139906.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P331809 | BC145788.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P331809 | BC145148.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P331809 | NM_013774.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P331809 | AF195492.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P331809 | AK139906.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2014626 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2112982 | BC145788.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2112982 | BC145148.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2112982 | NM_013774.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2112982 | AF195492.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2112982 | AK139906.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28707822 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28707837 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07736577 | - | - |
| Whole Mouse Genome G4122A | A_51_P331809 | BC145788.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P331809 | BC145148.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P331809 | NM_013774.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P331809 | AF195492.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P331809 | AK139906.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P331809 | BC145788.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P331809 | BC145148.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P331809 | NM_013774.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P331809 | AF195492.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P331809 | AK139906.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P130259 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28707894 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28707865 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28707926 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28707885 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28707931 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28707882 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28707927 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28707877 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28707915 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28707893 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28707891 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28707919 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28707895 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28707803 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28707880 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28707929 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28707889 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28707884 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28707910 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28707921 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28707886 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28707876 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28707924 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28707918 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10332861 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P09947030 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28707861 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28707837 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28707880 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28707884 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10388697 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28707885 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28707869 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10411371 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P09947035 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28707822 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28707864 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28707878 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28707882 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10332858 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28707858 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28707815 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10258019 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28707874 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10024198 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10398112 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10398114 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10398116 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10398115 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10398113 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1420423_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10398111 | - | - |
| Mouse Genome 430 2.0 Array | 1420423_at | - | - |
| Mouse Genome 430 2.0 Array | 1420423_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 27380_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 27380_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1200007558.mm.1 | NM_013774.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200007558.mm.1 | NM_013774.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200007558.mm.1 | TC1200001058.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1420423_PM_at | - | - |
| Mouse Expression Set 430 | 1420423_at | - | - |
| Mouse Genome 430A 2.0 Array | 1420423_at | - | - |
| Mouse Genome 430A 2.0 Array | 1420423_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1420423_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1420423_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 560055 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 6350685 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 6350685 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006350685 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1229633 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1229633 | - | - |
| mouse-6 v1.1 (Illumina) | scl42820.4.1_101-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6350685 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl42820.4.1_101-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6350685 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2697817 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2697817 | - | - |

Tcl1b4
Genomic Map