Assay Details
| Gene Symbol: Paox | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153783.4 | NP_722478.2 | 3 - 4 | 885 | 82 | |
| GenBank mRNA | AF226656.2 | - | 3 - 4 | 904 | 82 | |
| AK010469.1 | - | 2 - 3 | 534 | 83 | ||
| AK030664.1 | - | 3 - 4 | 886 | 82 | ||
| AK186235.1 | - | 1 - 2 | 39 | 82 | ||
| AM049402.1 | - | 3 - 4 | 877 | 82 | ||
| BC082783.1 | - | 3 - 4 | 884 | 6475809 | 82 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Paox
Genomic Map