Assay Details
| Gene Symbol: Klk1b9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010116.1 | NP_034246.1 | 243 | 137 | ||
| GenBank mRNA | BC024624.1 | - | 222 | 4914236 | 137 | |
| BC048869.1 | - | 207 | 4164261 | 137 | ||
| M17962.1 | - | 243 | 137 | |||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P221753 | BC024624.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P221753 | BC048869.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P221753 | M17962.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P221753 | NM_010116.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P221753 | BC024624.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P221753 | BC048869.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P221753 | M17962.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P221753 | NM_010116.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 170660 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 141138 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P221753 | BC024624.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P221753 | BC048869.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P221753 | M17962.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P221753 | NM_010116.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P221753 | BC024624.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P221753 | BC048869.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P221753 | M17962.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P221753 | NM_010116.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P221753 | BC024624.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P221753 | BC048869.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P221753 | M17962.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P221753 | NM_010116.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P221753 | BC024624.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P221753 | BC048869.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P221753 | M17962.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P221753 | NM_010116.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25085059 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25085070 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25085055 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25085048 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25085069 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25085041 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25085036 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25085035 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25085039 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06178810 | - | - |
| Whole Mouse Genome G4122A | A_51_P221753 | BC024624.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P221753 | BC048869.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P221753 | M17962.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P221753 | NM_010116.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P221753 | BC024624.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P221753 | BC048869.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P221753 | M17962.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P221753 | NM_010116.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P06178810 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06238713 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06196678 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25085103 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25085137 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25085081 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25085085 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25085114 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25085104 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25085110 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25085051 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25085004 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25085069 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25085095 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25085000 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25085036 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25085097 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25085087 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25085132 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25085035 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25085001 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25085019 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25085041 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25085018 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25085041 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25085085 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25085110 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25085048 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25085057 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25085051 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25085064 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06659858 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25085036 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25085097 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25085067 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06333310 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06612357 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25085111 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25085059 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25085080 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25085039 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06285663 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06429098 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25085104 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25085095 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25085103 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25085081 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06333321 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25085055 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25085106 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25085069 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10552568 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10552567 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10552569 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10552566 | - | - |
| Mouse Genome 430 2.0 Array | 1425182_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1425182_PM_x_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0700006367.mm.1 | NM_010116.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700006367.mm.1 | NM_010116.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700006367.mm.1 | TC0700000785.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700006368.mm.1 | NM_010116.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700006368.mm.1 | NM_010116.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700006368.mm.1 | TC0700000785.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700006370.mm.1 | NM_010116.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700006370.mm.1 | NM_010116.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700006370.mm.1 | TC0700000785.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700006369.mm.1 | NM_010116.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700006369.mm.1 | NM_010116.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700006369.mm.1 | TC0700000785.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1425182_PM_x_at | - | - |
| Mouse Expression Set 430 | 1425182_x_at | - | - |
| Murine Genome U74v2 Set | 162305_PM_f_at | - | - |
| Murine Genome U74v2 Set | 162305_f_at | - | - |
| Murine Genome U74v2 Set | 94716_PM_f_at | - | - |
| Murine Genome U74v2 Set | 94716_f_at | - | - |
| Mouse Genome 430A 2.0 Array | 1425182_x_at | - | - |
| Mouse Genome 430A 2.0 Array | 1425182_PM_x_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 396626 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 102060070 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 4120164 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004120164 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2723589 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1254219 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2723589 | - | - |
| mouse-6 v1.1 (Illumina) | GI_21426850-S | - | - |
| mouse-6 v1.1 (Illumina) | scl32731.3.1_44-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 102060070 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4120164 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl32731.3.1_44-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 102060070 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4120164 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2784773 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2723594 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2784773 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2723594 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1254219 | - | - |

Klk1b9
Genomic Map