Assay Details
| Gene Symbol: Henmt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001078646.1 | NP_001072114.1 | 7 - 8 | 905 | 141 | |
| NM_025723.2 | NP_079999.2 | 7 - 8 | 1220 | 141 | ||
| GenBank mRNA | AK038994.1 | - | 7 - 8 | 906 | 141 | |
| AK085587.1 | - | 7 - 8 | 1220 | 141 | ||
| BC099408.1 | - | 6 - 7 | 886 | 30935249 | 141 | |
| BC116412.1 | - | 5 - 6 | 579 | 141 | ||
| BC116413.1 | - | 5 - 6 | 701 | 141 | ||
| BC128494.1 | - | 5 - 6 | 701 | 141 | ||
| BC128495.1 | - | 5 - 6 | 701 | 141 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22415392 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22415393 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22415394 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2079957 | BC116413.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2079957 | BC128494.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2079957 | BC128495.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2079957 | AK038994.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2079957 | AK085587.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2079952 | AK038994.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2079952 | BC099408.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2079952 | NM_001078646.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2079952 | NM_025723.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 484183 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 547106 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 10785 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 7968 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 72584 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 465666 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 589808 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 881628 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 966550 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 617869 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 101835 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 149297 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 130609 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P145691 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P155838 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2079952 | AK038994.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2079952 | BC099408.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2079952 | NM_001078646.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2079952 | NM_025723.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2079957 | BC116413.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2079957 | BC128494.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2079957 | BC128495.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2079957 | AK038994.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2079957 | AK085587.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22415396 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22415408 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22415377 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22415395 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22415413 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22415387 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22415375 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22415381 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22415410 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22415389 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22415404 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22415407 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22415384 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22415397 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22415376 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22415380 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22415400 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22415382 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22415392 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22415411 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22415391 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22415406 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22415394 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22415402 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P145691 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P155838 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05026499 | - | - |
| Whole Mouse Genome G4122A | A_52_P284840 | AK038994.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P284840 | AK085587.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P284840 | BC099408.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P284840 | NM_001078646.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P284840 | NM_025723.2 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P140068 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P145691 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P155838 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02796476 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02796477 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03004217 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03089499 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02796473 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03004215 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22415393 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22415392 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22415394 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22415535 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22415504 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22415416 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22415486 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22415370 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22415501 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22415490 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22415510 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22415477 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22415451 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22415434 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22415396 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22415455 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22415468 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22415513 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22415413 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22415533 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22415432 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22415523 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22415362 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22415473 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22415511 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22415427 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22415525 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22415384 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22415392 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22415435 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22415454 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22415377 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22415465 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22415372 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22415453 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22415491 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22415457 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22415498 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22415472 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22415470 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22415466 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22415382 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22415519 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22415484 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22415376 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22415441 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22415409 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22415479 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22415509 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22415449 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22415381 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22415476 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22415407 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22415387 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22415471 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22415505 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22415534 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22415480 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22415474 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22415418 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22415411 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22415395 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22415492 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22415506 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22415485 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22415515 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22415482 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22415436 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22415400 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22415531 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22415420 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22415460 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22415375 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22415489 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22415415 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22415496 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22415367 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22415514 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22415537 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22415494 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22415402 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22415394 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22415430 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22415408 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22415371 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22415385 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03004217 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03089499 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22415418 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03068308 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22415408 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22415402 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22415415 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02296496 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22415398 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02296494 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02868853 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22415413 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22415372 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02796477 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22415377 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22415535 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02296488 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22415404 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22415381 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22415373 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22415400 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22415409 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22415533 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22415393 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22415534 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22415407 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22415387 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22415375 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02347651 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22415411 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22415396 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22415370 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10495391 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10495392 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10495386 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10495388 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10495393 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10495389 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10495387 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10495390 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10501455 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1430722_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10495385 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10501454 | - | - |
| Mouse Genome 430 2.0 Array | 1430722_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1430722_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 66715_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 66715_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0300009465.mm.1 | NM_001078646.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300009465.mm.1 | NM_001078646.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300009465.mm.1 | NM_025723.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300009465.mm.1 | NM_025723.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300009465.mm.1 | TC0300001156.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0300009466.mm.1 | NM_001078646.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300009466.mm.1 | NM_001078646.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300009466.mm.1 | NM_025723.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300009466.mm.1 | NM_025723.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300009466.mm.1 | TC0300001156.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1430722_at | - | - |
| Mouse Expression Set 430 | 1430722_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1430722_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1430722_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 835869 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 6900180 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006900180 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1218166 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1218166 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1215270 | - | - |
| mouse-6 v1.1 (Illumina) | scl22714.9.1_111-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6900180 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl22714.9.1_111-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6900180 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1218166 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_3111540 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1218166 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3111540 | - | - |

Henmt1
Genomic Map