Assay Details
| Gene Symbol: Mrpl9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030116.2 | NP_084392.1 | 4 - 5 | 490 | 116 | |
| GenBank mRNA | AB049637.1 | - | 4 - 5 | 450 | 116 | |
| AK021196.1 | - | 4 - 5 | 509 | 116 | ||
| AK040055.1 | - | 4 - 5 | 491 | 116 | ||
| AK046099.1 | - | 4 - 5 | 490 | 116 | ||
| AK146237.1 | - | 491 | 116 | |||
| AK161563.1 | - | 4 - 5 | 494 | 116 | ||
| AK162023.1 | - | 4 - 5 | 490 | 116 | ||
| BC116385.1 | - | 4 - 5 | 490 | 40101776 | 116 | |
| BC116386.1 | - | 4 - 5 | 490 | 40101778 | 116 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22339050 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22339052 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22339049 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22339051 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22339048 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2103541 | AK146237.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P136372 | AK162023.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P136372 | AK040055.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P136372 | AK046099.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P136372 | AK161563.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P136372 | NM_030116.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 583736 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 614188 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 151626 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P124220 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2103541 | AK146237.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P136372 | AK162023.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P136372 | AK040055.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P136372 | AK046099.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P136372 | AK161563.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P136372 | NM_030116.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P273130 | AK162023.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P273130 | BC116385.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P273130 | BC116386.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P273130 | AK046099.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P273130 | AK146237.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22339048 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22339052 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22339058 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22339053 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22339050 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22339054 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22339056 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22339060 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22339063 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22339051 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04992594 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P124220 | - | - |
| Whole Mouse Genome G4122A | A_51_P273130 | AK162023.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P273130 | BC116385.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P273130 | BC116386.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P273130 | AK046099.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P273130 | AK146237.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P136372 | AK162023.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P136372 | AK040055.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P136372 | AK046099.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P136372 | AK161563.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P136372 | NM_030116.2 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P124220 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02301655 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22339049 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22339052 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22339048 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22339051 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22339050 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22339058 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22339014 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22339080 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22339003 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22339078 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22339027 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22339012 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22339045 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22339023 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22339052 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22339038 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22339079 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22339051 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22339069 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22339042 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22339121 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22339072 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22339092 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22338999 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22339008 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22339061 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22339086 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22339004 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22339005 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22339056 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22339114 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22339037 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22339075 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22339016 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22339035 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22339090 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22339011 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22339050 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22339046 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22339070 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22339082 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22339125 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22339095 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22339007 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22339030 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22339116 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22339112 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22339021 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22339002 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22339047 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22339065 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22339063 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22339087 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22339033 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22339048 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22339122 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22339117 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22339028 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22339121 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22339040 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22339080 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02639751 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22339061 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22339038 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22339041 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22339122 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22339066 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22339069 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02655855 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22339037 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22339078 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22339055 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22339072 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22339033 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02792735 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02301657 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22339070 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22339065 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22339087 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22339125 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22339052 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22339075 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22339042 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22339050 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22339058 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22339054 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22339086 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22339045 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02655853 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22339067 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22339056 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22339088 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22339064 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02462714 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22339063 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02352733 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22339084 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22339053 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22339051 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10494066 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10494068 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10494062 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10494065 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10494061 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10494067 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10494064 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10494063 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1427297_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1430976_PM_a_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1437749_PM_s_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10494060 | - | - |
| Mouse Genome 430 2.0 Array | 1430976_PM_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1430976_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1437749_PM_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1427297_at | - | - |
| Mouse Genome 430 2.0 Array | 1437749_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1427297_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 78523_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 78523_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0300006649.mm.1 | NM_030116.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300006649.mm.1 | NM_030116.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300006649.mm.1 | TC0300000869.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0300006651.mm.1 | NM_030116.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300006651.mm.1 | NM_030116.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300006651.mm.1 | TC0300000869.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0300006646.mm.1 | NM_030116.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300006646.mm.1 | NM_030116.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300006646.mm.1 | TC0300000869.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0300006647.mm.1 | NM_030116.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300006647.mm.1 | NM_030116.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300006647.mm.1 | TC0300000869.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0300006652.mm.1 | NM_030116.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300006652.mm.1 | NM_030116.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300006652.mm.1 | TC0300000869.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1430976_a_at | - | - |
| Mouse Expression Set 430 | 1430976_PM_a_at | - | - |
| Mouse Expression Set 430 | 1427297_at | - | - |
| Mouse Expression Set 430 | 1427297_PM_at | - | - |
| Mouse Expression Set 430 | 1437749_s_at | - | - |
| Mouse Expression Set 430 | 1437749_PM_s_at | - | - |
| Murine Genome U74v2 Set | 102058_PM_at | - | - |
| Murine Genome U74v2 Set | 102058_at | - | - |
| Mouse Genome 430A 2.0 Array | 1430976_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1430976_PM_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1427297_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1427297_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1430976_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1427297_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1430976_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1427297_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1437749_s_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1437749_PM_s_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 341965 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 580079 | - | - |
| MouseWG-6 v2 Expression BeadChip | 103120369 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 580079 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 103120369 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0000580079 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2679770 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2679770 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2534975 | - | - |
| mouse-6 v1.1 (Illumina) | scl22902.7.1_30-S | - | - |
| mouse-6 v1.1 (Illumina) | GI_29789252-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 580079 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 103120369 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl22902.7.1_30-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 580079 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 103120369 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2679770 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2534975 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2679770 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl22902.7.1_30-S | - | - |

Mrpl9
Genomic Map