Assay Details
| Gene Symbol: Fam131a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133778.2 | NP_598539.1 | 5 - 6 | 654 | 102 | |
| GenBank mRNA | AK049983.1 | - | 5 - 6 | 655 | 102 | |
| BC092532.1 | - | 5 - 6 | 685 | 6594874 | 102 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30589002 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30589003 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30589004 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30589023 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30589024 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30589022 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30589000 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2031481 | BC003957.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2031481 | BC092532.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2031481 | NM_133778.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2031481 | AK019338.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2031481 | AK049983.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 175255 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 48357 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 590625 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 570846 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 27230 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P105761 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2031481 | BC003957.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2031481 | BC092532.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2031481 | NM_133778.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2031481 | AK019338.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2031481 | AK049983.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30589007 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30589015 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30589019 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30589004 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30589005 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30589001 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30589033 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30588998 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30589027 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30589024 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30589023 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30589017 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30589014 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30589012 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30589009 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30589010 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30589020 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30589026 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30589021 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30589028 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30589000 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30589025 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30589002 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30589031 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08527791 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P105761 | - | - |
| Whole Mouse Genome G4122A | A_51_P213205 | BC003957.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P213205 | BC092532.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P213205 | NM_133778.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P213205 | AK019338.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P213205 | AK049983.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P133691 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P105761 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12541531 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12863653 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12863662 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12507686 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12863656 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12863661 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30589023 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30589022 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30589004 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30589024 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30589002 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30589003 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30589000 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30589146 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30589103 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30589065 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30589133 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30589000 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30589046 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30589020 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30589047 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30589137 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30589037 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30589062 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30589002 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30589045 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30589121 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30589109 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30589118 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30589130 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30589056 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30589027 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30589042 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30589051 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30589129 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30589097 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30589120 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30589012 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30589113 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30589023 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30589033 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30589105 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30589071 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30589139 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30589015 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30589007 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30589038 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30589019 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30589132 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30589069 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30589108 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30589102 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30589112 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30589024 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30589052 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30589055 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30589142 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30589147 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30589010 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30589017 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30589070 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30589025 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30589039 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30589001 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30589123 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30589138 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30589093 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30589014 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30588998 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30589061 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30589106 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30589067 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30589005 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30589125 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30589049 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30589110 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30589028 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30589115 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30589100 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30589064 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30589004 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30589063 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30589128 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30589027 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30589051 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30589002 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12541531 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30589062 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12507696 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30589012 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12881267 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30589019 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30589052 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12709766 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12546592 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30589065 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30589017 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12727276 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30589020 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30589014 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30589041 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30589039 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30589067 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30589026 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30589047 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30589055 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30589024 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30589036 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30589004 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30589046 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30589069 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30589063 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30589057 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30589042 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30589007 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30589033 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30588998 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30589005 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30589031 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30589058 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30589028 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30589037 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12546595 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12881262 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30589045 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12863661 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30589056 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10434522 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10434517 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10434521 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10434519 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10434518 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10434520 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1426729_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10434516 | - | - |
| Mouse Genome 430 2.0 Array | 1426729_at | - | - |
| Mouse Genome 430 2.0 Array | 1426729_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 78408_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 78408_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1600002878.mm.1 | NM_133778.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600002878.mm.1 | NM_133778.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600002878.mm.1 | TC1600000290.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1600002879.mm.1 | NM_133778.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600002879.mm.1 | NM_133778.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600002879.mm.1 | TC1600000290.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1600002877.mm.1 | NM_133778.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600002877.mm.1 | NM_133778.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600002877.mm.1 | TC1600000290.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1426729_PM_at | - | - |
| Mouse Expression Set 430 | 1426729_at | - | - |
| Murine Genome U74v2 Set | 165462_at | - | - |
| Murine Genome U74v2 Set | 165462_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1426729_at | - | - |
| Mouse Genome 430A 2.0 Array | 1426729_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1426729_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1426729_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 553360 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 6840167 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 6840167 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006840167 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2713060 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2713060 | - | - |
| mouse-6 v1.1 (Illumina) | scl078408.1_245-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6840167 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl078408.1_245-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6840167 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2713060 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2713060 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl078408.1_245-S | - | - |

Fam131a
Genomic Map