Assay Details
| Gene Symbol: Chpf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133913.2 | NP_598674.2 | 3 - 4 | 2298 | 61 | |
| GenBank mRNA | AK089404.1 | - | 3 - 4 | 2308 | 61 | |
| AK122504.1 | - | 2 - 3 | 2945 | 61 | ||
| AK138814.1 | - | 2304 | 61 | |||
| BC019714.1 | - | 2 - 3 | 359 | 61 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_65_P15875 | AK138814.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_65_P15875 | BC019714.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_65_P15875 | BC036970.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_65_P15875 | BC047275.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_65_P15875 | AK122504.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1987904 | AK138814.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1987912 | AK138814.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1987912 | BC019714.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1987912 | BC036970.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1987912 | BC047275.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1987912 | AK122504.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 552356 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 157725 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 741666 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 47089 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P147057 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1987912 | AK138814.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1987912 | BC019714.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1987912 | BC036970.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1987912 | BC047275.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1987912 | AK122504.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_65_P15875 | AK138814.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_65_P15875 | BC019714.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_65_P15875 | BC036970.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_65_P15875 | BC047275.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_65_P15875 | AK122504.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P295389 | BC019714.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P479326 | AK138814.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P479326 | BC047275.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P479326 | AK122504.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P479326 | NM_133913.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P479326 | AK089404.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P125824 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23542868 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23542920 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23542863 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23542927 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23542974 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23542858 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23542918 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23542955 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23542937 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23542830 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23542891 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23542976 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23542940 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23542823 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23542942 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23542854 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23542967 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23542803 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23542816 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23542894 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23542871 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23542973 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23542935 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23542866 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23542958 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23542817 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23542930 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23542938 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23542956 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23542851 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23542878 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23542924 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23542831 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23542821 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23542812 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23542965 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23542885 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23542889 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23542948 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23542972 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23542843 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23542840 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23542944 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23542799 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23542880 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23542839 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23542807 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23542882 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23542837 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23542808 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23542877 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23542818 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23542931 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23542856 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23542876 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23542969 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23542941 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23542949 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23542968 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23542933 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23542978 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23542820 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23542950 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23542804 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23542890 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23542801 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23542963 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23542873 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23542825 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23542916 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23542875 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23542953 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23542887 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23542922 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23542946 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23542848 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23542853 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23542971 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23542893 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23542977 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04840537 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23542858 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23542841 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04518915 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23542863 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04355377 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23542887 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04355396 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04840552 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04840557 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04840530 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23542837 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04355365 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04555603 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04840543 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04355398 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04840551 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04355388 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23542840 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04840563 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23542839 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23542849 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23542829 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23542875 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04840527 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04840525 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04518916 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23542894 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04355400 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04355381 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04355364 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04355379 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04355374 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04355403 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04355370 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23542893 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04921870 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04840528 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04840535 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10520238 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10520241 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10520244 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10520246 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10520239 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10520243 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10520240 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10520245 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10520235 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10520242 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10520236 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1453846_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1452170_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10520234 | - | - |
| Mouse Genome 430 2.0 Array | 1452170_at | - | - |
| Mouse Genome 430 2.0 Array | 1452170_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1444415_at | - | - |
| Mouse Genome 430 2.0 Array | 1444415_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0500001685.mm.1 | NM_133913.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500001685.mm.1 | NM_133913.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500001685.mm.1 | NR_030423.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500001685.mm.1 | NR_030423.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500001685.mm.1 | TC0500000219.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500001684.mm.1 | NM_133913.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500001684.mm.1 | NM_133913.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500001684.mm.1 | TC0500000219.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500001686.mm.1 | NM_133913.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500001686.mm.1 | NM_133913.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500001686.mm.1 | TC0500000219.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500001682.mm.1 | NM_133913.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500001682.mm.1 | NM_133913.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500001682.mm.1 | TC0500000219.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500001683.mm.1 | NM_133913.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500001683.mm.1 | NM_133913.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500001683.mm.1 | TC0500000219.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1452170_at | - | - |
| Mouse Expression Set 430 | 1452170_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1452170_at | - | - |
| Mouse Genome 430A 2.0 Array | 1452170_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1452170_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1452170_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 669874 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 3840110 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0003840110 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2702868 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2702868 | - | - |
| mouse-6 v1.1 (Illumina) | scl00100910.1_246-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 3840110 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl00100910.1_246-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3840110 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2702868 | - | - |

Chpf2
Genomic Map