Assay Details
| Gene Symbol: Lrpap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013587.2 | NP_038615.2 | 1 - 2 | 221 | 72 | |
| GenBank mRNA | AJ639614.1 | - | 1 - 2 | 228 | 72 | |
| AK089250.1 | - | 1 - 2 | 224 | 72 | ||
| AK145710.1 | - | 1 - 2 | 264 | 72 | ||
| AK150451.1 | - | 1 - 2 | 235 | 72 | ||
| AK152485.1 | - | 1 - 2 | 235 | 72 | ||
| AK153323.1 | - | 1 - 2 | 235 | 72 | ||
| AK166616.1 | - | 1 - 2 | 224 | 72 | ||
| AK166746.1 | - | 1 - 2 | 235 | 72 | ||
| AK166819.1 | - | 1 - 2 | 239 | 72 | ||
| AK168421.1 | - | 1 - 2 | 224 | 72 | ||
| AK169132.1 | - | 1 - 2 | 229 | 72 | ||
| AK171471.1 | - | 1 - 2 | 224 | 72 | ||
| BC032170.1 | - | 1 - 2 | 224 | 72 | ||
| BC046641.1 | - | 1 - 2 | 213 | 72 | ||
| BC057979.1 | - | 1 - 2 | 192 | 72 | ||
| BC094324.1 | - | 1 - 2 | 221 | 6402163 | 72 | |
| D00622.1 | - | 1 - 2 | 214 | 72 | ||
| S67967.1 | - | 1 - 2 | 247 | 72 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Lrpap1
Genomic Map